2-Amino-3-ethyl-6-methylpyridine, 97%, Thermo Scientific Chemicals
2-Amino-3-ethyl-6-methylpyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-3-ethyl-6-methylpyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A19048.06
also known as A19048-06
Price (USD)/ Each
80.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
80.90
Add to cart
2-Amino-3-ethyl-6-methylpyridine, 97%, Thermo Scientific Chemicals
Catalog numberA19048.06
Price (USD)/ Each
80.90
-
Add to cart
Chemical Identifiers
CAS41995-31-7
IUPAC Name2-amino-3-ethyl-6-methylpyridin-1-ium
Molecular FormulaC8H13N2
InChI KeyOQNDNGDJFBXUID-UHFFFAOYSA-O
SMILESCCC1=CC=C(C)[NH+]=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Assay (GC)≥96.0%
FormPowder
Melting Point (clear melt)43.0-49.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only