N-Benzyloxycarbonyl-L-serine, 99%, Thermo Scientific Chemicals
N-Benzyloxycarbonyl-L-serine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-serine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A19050.22
also known as A19050-22
Price (USD)/ Each
298.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
298.00
Add to cart
N-Benzyloxycarbonyl-L-serine, 99%, Thermo Scientific Chemicals
Catalog numberA19050.22
Price (USD)/ Each
298.00
-
Add to cart
Chemical Identifiers
CAS1145-80-8
IUPAC Name2-{[(benzyloxy)carbonyl]amino}-3-hydroxypropanoic acid
Molecular FormulaC11H13NO5
InChI KeyGNIDSOFZAKMQAO-UHFFFAOYNA-N
SMILESOCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormPowder
Melting Point (clear melt)115.0-121.0?C
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only