Reaction of BunLi with trithiocyanuric acid (LH3) in toluene/HMPA yielded a mono-lithiated species, [(LH2Li)·HMPA2]. Metal complexes of trithiocyanuric acid is used for the elucidation of coordination polymer stoichiometry via thermometric titrimetry:.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reaction of BunLi with trithiocyanuric acid (LH3) in toluene/HMPA yielded a mono-lithiated species, [(LH2Li)·HMPA2]. Metal complexes of trithiocyanuric acid is used for the elucidation of coordination polymer stoichiometry via thermometric titrimetry:.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
- William J. Hunks,; Michael C. Jennings,; Richard J. Puddephatt. Gold Complexes of Trithiocyanuric Acid: A Two-Dimensional Polymer Assembled through Gold(I)···Gold(I) Interactions. Inorg. Chem. 1999, 38 (26), 5930-5931.
- A.E. Beezer,; J.C. Chudy.Elucidation of coordination polymer stoichiometry via thermometric titrimetry: Metal complexes of trithiocyanuric acid. Thermochimica Acta. 1973, 6 (2), 231-237.