Thermo Scientific Chemicals

Bromocresol Purple sodium salt

Catalog number: A19082.18
50 g, Each
Thermo Scientific Chemicals

Bromocresol Purple sodium salt

Catalog number: A19082.18
50 g, Each
Quantity
Catalog number: A19082.18
also known as A19082-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62625-30-3
IUPAC Name
2-[(3-bromo-4-hydroxy-5-methylphenyl)[(1E)-3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
Molecular Formula
C21H15Br2O5S
InChI Key
HJZFRHKAXRLQSG-XMHGGMMESA-M
SMILES
CC1=CC(=CC(Br)=C1O)C(=C1\C=C(C)C(=O)C(Br)=C1)\C1=CC=CC=C1S([O-])(=O)=O
lambda-max (UV-Vis)
589±3nm (0.01M NaOH)
Form
Powder
Color Change
Yellow @ pH5.2 to Purple @ pH6.8
Specific Absorbance (E 1%/1cm)
≥850 (lambda max)
Appearance (Color)
Orange to red or brown

Description

Bromocresol purple sodium salt is used to isolate acid-producing bacteria from sauerkraut juice, and lactic bacteria is scalped by morphology and share, then morphological, physiological and biochemical interactions. It can be used as a fluorescent stain to detect yeast cells with plasma membrane damage for dead cell counts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocresol purple sodium salt is used to isolate acid-producing bacteria from sauerkraut juice, and lactic bacteria is scalped by morphology and share, then morphological, physiological and biochemical interactions. It can be used as a fluorescent stain to detect yeast cells with plasma membrane damage for dead cell counts.

Solubility
Soluble in water.

Notes
Transition interval: pH 5.2 (yellow) to pH 6.8 (purple)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text