Thermo Scientific Chemicals

2-Methyl-2-pentanol, 99%, Thermo Scientific Chemicals

Catalog number: A19090.14
25 g, Each
Thermo Scientific Chemicals

2-Methyl-2-pentanol, 99%, Thermo Scientific Chemicals

Catalog number: A19090.14
25 g, Each
Quantity
Catalog number: A19090.14
also known as A19090-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
590-36-3
IUPAC Name
2-methylpentan-2-ol
Molecular Formula
C6H14O
InChI Key
WFRBDWRZVBPBDO-UHFFFAOYSA-N
SMILES
CCCC(C)(C)O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.4085-1.4125 @ 20?C

Description

It is used to study its antagonist activity. It reacts with a second target site to alter the spatial relation between the hydrophobic agonist binding site and the allosteric site. It may initiate the polymerization of isocyanates and epoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to study its antagonist activity. It reacts with a second target site to alter the spatial relation between the hydrophobic agonist binding site and the allosteric site. It may initiate the polymerization of isocyanates and epoxides.

Solubility
Soluble in water at 20°C 32g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text