Thermo Scientific Chemicals

Spermidine, 99%, Thermo Scientific Chemicals

Catalog number: A19096.06
5 g, Each
Thermo Scientific Chemicals

Spermidine, 99%, Thermo Scientific Chemicals

Catalog number: A19096.06
5 g, Each
Quantity
Catalog number: A19096.06
also known as A19096-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-20-9
Assay (GC)
≥98.5% (non-U.S. specification)
Appearance (Color)
Clear colorless to pale yellow or white to pale cream
Assay from Suppliers CofA
≥98.5% (U.S. specification : GC)
Refractive Index
1.4770-1.4810 @ 20°C (non-U.S. specification)
Form
Liquid as melt or solid @ <20°C

Description

Binds to the polyamine modulatory site of NMDA Spermidine is used in electroporation during transferring the DNA into the cell under the electrical impulse. It is also used for purification of DNA-binding proteins. It combines with calcium chloride and utilized for the precipitation of DNA onto microprojectiles for bombardment with a gene gun. It plays an important role to reduce the amount of aging in yeast, flies, worms, and human immune cells by inducing autophagy. It serves as a precursor of spermine and essential for both normal and neoplastic tissue growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Binds to the polyamine modulatory site of NMDA Spermidine is used in electroporation during transferring the DNA into the cell under the electrical impulse. It is also used for purification of DNA-binding proteins. It combines with calcium chloride and utilized for the precipitation of DNA onto microprojectiles for bombardment with a gene gun. It plays an important role to reduce the amount of aging in yeast, flies, worms, and human immune cells by inducing autophagy. It serves as a precursor of spermine and essential for both normal and neoplastic tissue growth.

Solubility
Miscible with water, ethanol and ether.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text