3-Nonanol, 98%, Thermo Scientific Chemicals
3-Nonanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Nonanol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number A19120.30
also known as A19120-30
Price (USD)/ Each
622.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
622.00
Add to cart
3-Nonanol, 98%, Thermo Scientific Chemicals
Catalog numberA19120.30
Price (USD)/ Each
622.00
-
Add to cart
Chemical Identifiers
CAS624-51-1
IUPAC Namenonan-3-ol
Molecular FormulaC9H20O
InChI KeyGYSCXPVAKHVAAY-UHFFFAOYNA-N
SMILESCCCCCCC(O)CC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4280-1.4320 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only