4-Bromo-3-fluorotoluene, 98%, Thermo Scientific Chemicals
4-Bromo-3-fluorotoluene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-3-fluorotoluene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Catalog number A19142.09
also known as A19142-09
Price (USD)/ Each
103.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
103.00
Add to cart
4-Bromo-3-fluorotoluene, 98%, Thermo Scientific Chemicals
Catalog numberA19142.09
Price (USD)/ Each
103.00
-
Add to cart
Chemical Identifiers
CAS452-74-4
IUPAC Name1-bromo-2-fluoro-4-methylbenzene
Molecular FormulaC7H6BrF
InChI KeySLFNGVGRINFJLK-UHFFFAOYSA-N
SMILESCC1=CC(F)=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5290-1.5330 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only