Thermo Scientific Chemicals

4-Bromo-3-fluorotoluene, 98%, Thermo Scientific Chemicals

Catalog number: A19142.14
25 g, Each
Thermo Scientific Chemicals

4-Bromo-3-fluorotoluene, 98%, Thermo Scientific Chemicals

Catalog number: A19142.14
25 g, Each
Quantity
Catalog number: A19142.14
also known as A19142-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
452-74-4
IUPAC Name
1-bromo-2-fluoro-4-methylbenzene
Molecular Formula
C7H6BrF
InChI Key
SLFNGVGRINFJLK-UHFFFAOYSA-N
SMILES
CC1=CC(F)=C(Br)C=C1
Form
Liquid
Refractive Index
1.5290-1.5330 @ 20?C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text