Thermo Scientific Chemicals

cis-2-Butene-1,4-diol, 96%, remainder trans-isomer, Thermo Scientific Chemicals

Catalog number: A19182.0B
1000 g, Each
Thermo Scientific Chemicals

cis-2-Butene-1,4-diol, 96%, remainder trans-isomer, Thermo Scientific Chemicals

Catalog number: A19182.0B
1000 g, Each
Quantity
Catalog number: A19182.0B
also known as A19182-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6117-80-2
IUPAC Name
(2Z)-but-2-ene-1,4-diol
Molecular Formula
C4H8O2
InChI Key
ORTVZLZNOYNASJ-UPHRSURJSA-N
SMILES
OC\C=C/CO
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥95.0%
Refractive Index
1.4750-1.4810 @ 20?C

Description

cis-2-Butene-1, 4-diol is used as a probe for studying isomerization versus hydrogenation and hydrogenolysis reactions. (DPCB-OMe) efficiently catalyzes cyclodehydration of cis-2-butene-1,4-diol with active methylene compounds to give 2-vinyl-2,3-dihydrofurans in good to high yields. Grubbs's cross metathesis of eugenol with cis-2-butene-1, 4-diol to make a natural product, an organometallic experiment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-2-Butene-1, 4-diol is used as a probe for studying isomerization versus hydrogenation and hydrogenolysis reactions. (DPCB-OMe) efficiently catalyzes cyclodehydration of cis-2-butene-1,4-diol with active methylene compounds to give 2-vinyl-2,3-dihydrofurans in good to high yields. Grubbs′s cross metathesis of eugenol with cis-2-butene-1, 4-diol to make a natural product, an organometallic experiment.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text