Thermo Scientific Chemicals

Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A19188.06
5 g, Each
Thermo Scientific Chemicals

Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A19188.06
5 g, Each
Quantity
Catalog number: A19188.06
also known as A19188-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
593-56-6
IUPAC Name
hydrogen O-methylhydroxylamine chloride
Molecular Formula
CH6ClNO
InChI Key
XNXVOSBNFZWHBV-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CON
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Form
Crystals or powder or crystalline powder or fused solid

Description

Methoxylamine hydrochloride is used as a reagent in the preparation of O-methyl oximes from aldehydes. It is orally bioavailable small molecule inhibitor with potential adjuvant activity. It is also used as internal standard for prostaglandin assays by gas chromatography-mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxylamine hydrochloride is used as a reagent in the preparation of O-methyl oximes from aldehydes. It is orally bioavailable small molecule inhibitor with potential adjuvant activity. It is also used as internal standard for prostaglandin assays by gas chromatography-mass spectrometry.

Solubility
Soluble in water, dimethyl sulfoxide and methanol.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text