3,5-Dihydroxy-4-methylbenzoic acid, 97%, Thermo Scientific Chemicals
3,5-Dihydroxy-4-methylbenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dihydroxy-4-methylbenzoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A19196.06
also known as A19196-06
Price (USD)/ Each
162.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
162.00
Add to cart
3,5-Dihydroxy-4-methylbenzoic acid, 97%, Thermo Scientific Chemicals
Catalog numberA19196.06
Price (USD)/ Each
162.00
-
Add to cart
Chemical Identifiers
CAS28026-96-2
IUPAC Name3,5-dihydroxy-4-methylbenzoate
Molecular FormulaC8H7O4
InChI KeyKMRRXSZDSGYLCD-UHFFFAOYSA-M
SMILESCC1=C(O)C=C(C=C1O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)> 96.0%
Appearance (Color)White to pale cream
Formpowder
Assay (Silylated GC)> 96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only