Thermo Scientific™

tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals

Catalog number: A19206.0B
1000 g, Each
Thermo Scientific™

tert-Butylhydroquinone, 97%, Thermo Scientific Chemicals

Catalog number: A19206.0B
1000 g, Each
Quantity
Catalog number: A19206.0B
also known as A19206-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1948-33-0
IUPAC Name
2-tert-butylbenzene-1,4-diol
Molecular Formula
C10H14O2
InChI Key
BGNXCDMCOKJUMV-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC(O)=CC=C1O
Assay (GC)
≥96.0% (UK-sourced material)
Melting Point
123.0-132.0?C
Appearance (Color)
White to cream
Comment
Material sourced in UK and US
Form
Crystals or powder or crystalline powder

Description

TBHQ was used to study the inactivation of barotolerant strains of Listeria monocytogenes and Escherichia coli. Environment friendly electrode materials for supercapacitors were attained by decorating the surface of graphene nanosheets with TBHQ. TBHQ is frequently used as a food preservative. Inhibition of oxidative rancidity in frozen cooked fish flakes by tert-butylhydroquinone. Nuclear factor E2-related factor 2-dependent antioxidant response element activation is by tert-butylhydroquinoneoccurring preferentially in Astrocytes conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TBHQ was used to study the inactivation of barotolerant strains of Listeria monocytogenes and Escherichia coli. Environment friendly electrode materials for supercapacitors were attained by decorating the surface of graphene nanosheets with TBHQ. TBHQ is frequently used as a food preservative. Inhibition of oxidative rancidity in frozen cooked fish flakes by tert-butylhydroquinone. Nuclear factor E2-related factor 2-dependent antioxidant response element activation is by tert-butylhydroquinoneoccurring preferentially in Astrocytes conditions.

Solubility
Slightly soluble in water(10g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text