Thermo Scientific Chemicals

5-Chlorooxindole, 98%, Thermo Scientific Chemicals

Catalog number: A19210.06
5 g, Each
Thermo Scientific Chemicals

5-Chlorooxindole, 98%, Thermo Scientific Chemicals

Catalog number: A19210.06
5 g, Each
Quantity
Catalog number: A19210.06
also known as A19210-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17630-75-0
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream to pink to pale brown
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
194-201?C

Description

5-Chloro-2-oxindole was used for the quantitation of 5-chloro-2-oxindole, concomitantly with all of its potential positional isomers using a single, highly specific, normal-phase chromatographic system. 5-Chloro-2-oxindole (5-Chlorooxindole) is a starting material for tenidap sodium, a pharmaceutical drug candidate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-2-oxindole was used for the quantitation of 5-chloro-2-oxindole, concomitantly with all of its potential positional isomers using a single, highly specific, normal-phase chromatographic system. 5-Chloro-2-oxindole (5-Chlorooxindole) is a starting material for tenidap sodium, a pharmaceutical drug candidate.

Solubility
Slightly Soluble (2.0 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text