Decafluorobenzophenone, 97%, Thermo Scientific Chemicals
Decafluorobenzophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Decafluorobenzophenone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number A19262.06
also known as A19262-06
Price (USD)/ Each
163.65
Online exclusive
182.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
163.65
Online exclusive
182.00 
Save 18.35 (10%)
Add to cart
Decafluorobenzophenone, 97%, Thermo Scientific Chemicals
Catalog numberA19262.06
Price (USD)/ Each
163.65
Online exclusive
182.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS853-39-4
IUPAC Namebis(2,3,4,5,6-pentafluorophenyl)methanone
Molecular FormulaC13F10O
InChI KeyWWQLXRAKBJVNCC-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(C(=O)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)89.0-95.0°C
FormCrystals or powder or crystalline powder or lumps or fused solid
Appearance (Color)White
Assay (GC)≥96.0%
Perfluorobenzophenone was used in the synthesis of fluorinated acridones and acridines. Fluorinated polyetherketones prepared from decafluorobenzophenone. Laser pulse excitation of decafluorobenzophenone (DFB) in acetonitrile yields the triplet 3DFB.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluorobenzophenone was used in the synthesis of fluorinated acridones and acridines. Fluorinated polyetherketones prepared from decafluorobenzophenone. Laser pulse excitation of decafluorobenzophenone (DFB) in acetonitrile yields the triplet 3DFB.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Del Buttero P, et al. Orthogonal Synthesis of Fluorinated Acridones and Acridines from Perfluorobenzophenone. European J. Org. Chem. 2011, (12), 2265-2271.
  2. Manuel J. Aroney,; Kingsley E. Calderbank,; Raymond K. Pierens. Electric birefringences and solution-state conformations of decafluorobiphenyl and decafluorobenzophenone. Journal of Molecular Structure. 1985 , 127 (1-2), 175-180.