Thermo Scientific Chemicals

3,4,5-Trihydroxybenzaldehyde hydrate, 97%, Thermo Scientific Chemicals

Catalog number: A19270.06
5 g, Each
Thermo Scientific Chemicals

3,4,5-Trihydroxybenzaldehyde hydrate, 97%, Thermo Scientific Chemicals

Catalog number: A19270.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A19270.06
also known as A19270-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13677-79-7
IUPAC Name
3,4,5-trihydroxybenzaldehyde
Molecular Formula
C7H6O4
InChI Key
RGZHEOWNTDJLAQ-UHFFFAOYSA-N
SMILES
OC1=CC(C=O)=CC(O)=C1O
Assay (HPLC)
≥96.0%
Form
Crystals or powder or crystalline powder or needles
Water Content (Karl Fischer Titration)
≤22.61%
Appearance (Color)
Pale cream to cream to brown

Description

3,4,5-trihydroxybenzaldehyde from Geum japonicum has dual inhibitory effect on matrix metalloproteinase 9; inhibition of gelatinoytic activity as well as MMP-9 expression in TNF-α induced HASMC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-trihydroxybenzaldehyde from Geum japonicum has dual inhibitory effect on matrix metalloproteinase 9; inhibition of gelatinoytic activity as well as MMP-9 expression in TNF-α induced HASMC.

Solubility
Soluble in DMSO and Methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text