3,5-Bis(trifluoromethyl)benzyl alcohol, 98%, Thermo Scientific Chemicals
3,5-Bis(trifluoromethyl)benzyl alcohol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)benzyl alcohol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A19290.06
also known as A19290-06
Price (USD)/ Each
89.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
89.10
Add to cart
3,5-Bis(trifluoromethyl)benzyl alcohol, 98%, Thermo Scientific Chemicals
Catalog numberA19290.06
Price (USD)/ Each
89.10
-
Add to cart
Chemical Identifiers
CAS32707-89-4
IUPAC Name[3,5-bis(trifluoromethyl)phenyl]methanol
Molecular FormulaC9H6F6O
InChI KeyBJTWPJOGDWRYDD-UHFFFAOYSA-N
SMILESOCC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Appearance (Color)White
Melting Point (clear melt)52.0-58.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only