Thermo Scientific Chemicals

N,N'-Diisopropylcarbodiimide, 99%, Thermo Scientific Chemicals

Catalog number: A19292.30
250 g, Each
Thermo Scientific Chemicals

N,N'-Diisopropylcarbodiimide, 99%, Thermo Scientific Chemicals

Catalog number: A19292.30
250 g, Each
Quantity
Catalog number: A19292.30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
693-13-0
IUPAC Name
(propan-2-yl)({[(propan-2-yl)imino]methylidene})amine
Molecular Formula
C7H14N2
InChI Key
BDNKZNFMNDZQMI-UHFFFAOYSA-N
SMILES
CC(C)N=C=NC(C)C
Form
Liquid
Comment
Material Sourced in UK and US
Refractive Index
1.4320-1.4350 @ 20?C (UK Sourced Material)
Appearance (Color)
Clear colorless to yellow
Assay from Supplier's CofA
≥98.5% (US Sourced Material)

Description

N,N'-Diisopropylcarbodiimide is used as a reagent in synthetic organic chemistry. It serves as a chemical intermediate and as a stabilizer for Sarin (chemical weapon). It is also used in the synthesis of peptide and nucleic acid. Further, it is used as an antineoplastic and involved in the treatment of malignant melanoma and sarcomas. In addition to this, it is used in the synthesis of acid anhydride, aldehyde, ketone and isocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diisopropylcarbodiimide is used as a reagent in synthetic organic chemistry. It serves as a chemical intermediate and as a stabilizer for Sarin (chemical weapon). It is also used in the synthesis of peptide and nucleic acid. Further, it is used as an antineoplastic and involved in the treatment of malignant melanoma and sarcomas. In addition to this, it is used in the synthesis of acid anhydride, aldehyde, ketone and isocyanate.

Solubility
Soluble in chloroform, methylene chloride, acetonitrile, dioxane, dimethylformamide andtetrahydrofuran.

Notes
Moisture sensitive. Store in a cool place Incompatible with oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text