2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.
Solubility
Immiscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only
General References:
- McInroy, G. R.; Raiber, E. A.; Balasubramanian, S. Chemical biology of genomic DNA: minimizing PCR bias. Chem. Commun. 2014, 50, 12047-12049.
- Gao, F.; Deng, X. J.; Tang, Y.; Tang, J. P.; Yang, J.; Zhang, Y. M. A simple and efficient copper oxide-catalyzed Barbier-Grignard reaction of unactivated aryl or alkyl bromides with ester. Tetrahedron Lett. 2014, 55 (4), 880-883.