Thermo Scientific Chemicals

2-Bromomethyl-1,3-dioxolane, 97%, Thermo Scientific Chemicals

Catalog number: A19312.36
500 g, Each
Thermo Scientific Chemicals

2-Bromomethyl-1,3-dioxolane, 97%, Thermo Scientific Chemicals

Catalog number: A19312.36
500 g, Each
Quantity
Catalog number: A19312.36
also known as A19312-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4360-63-8
IUPAC Name
2-(bromomethyl)-1,3-dioxolane
Molecular Formula
C4H7BrO2
InChI Key
CKIIJIDEWWXQEA-UHFFFAOYSA-N
SMILES
BrCC1OCCO1
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.4800-1.4850 @ 20?C

Description

2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromomethyl-1,3-dioxolane is used in the preparation of alfa, beta-unsaturated aldehyde and 1, 4-two aldehyde monoacetals synthesis of monomers.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text