Pyridine hydrobromide may be used in chemical synthesis studies. Pyridine hydrobromide is quite useful for brominations of ketones on a micro or semimicro scale.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pyridine hydrobromide may be used in chemical synthesis studies. Pyridine hydrobromide is quite useful for brominations of ketones on a micro or semimicro scale.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only
Carl Djerassi,; Caesar R. Scholz. Brominations with Pyridine Hydrobromide Perbromide. J. Am. Chem. Soc. 1984, 70 (1), 417-418.
Krishna C. Majumdar,; Anup K. Kundu. Pyridine hydrobromide perbromide mediated regioselective heterocyclization of ortho-cyclohexenyl phenols. JCanadian Journal of Chemistry. 1995, 73(10),1727-1732.