Tri-n-butyl borate, 98%, Thermo Scientific Chemicals
Tri-n-butyl borate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tri-n-butyl borate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number A19322.AE
also known as A19322-AE
Price (USD)/ Each
58.60
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
58.60
Add to cart
Tri-n-butyl borate, 98%, Thermo Scientific Chemicals
Catalog numberA19322.AE
Price (USD)/ Each
58.60
-
Add to cart
Chemical Identifiers
CAS688-74-4
IUPAC Nametributyl borate
Molecular FormulaC12H27BO3
InChI KeyLGQXXHMEBUOXRP-UHFFFAOYSA-N
SMILESCCCCOB(OCCCC)OCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
FormLiquid
Refractive Index1.4075-1.4115 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only