Thermo Scientific Chemicals

Tri-n-butyl borate, 98%, Thermo Scientific Chemicals

Catalog number: A19322.AP
500 mL, Each
Thermo Scientific Chemicals

Tri-n-butyl borate, 98%, Thermo Scientific Chemicals

Catalog number: A19322.AP
500 mL, Each
Quantity
Catalog number: A19322.AP
also known as A19322-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
688-74-4
IUPAC Name
tributyl borate
Molecular Formula
C12H27BO3
InChI Key
LGQXXHMEBUOXRP-UHFFFAOYSA-N
SMILES
CCCCOB(OCCCC)OCCCC
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.4075-1.4115 @ 20?C
Assay (GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text