Thermo Scientific Chemicals

Pentafluoroaniline, 97%, Thermo Scientific Chemicals

Catalog number: A19348.06
5 g, Each
Thermo Scientific Chemicals

Pentafluoroaniline, 97%, Thermo Scientific Chemicals

Catalog number: A19348.06
5 g, Each
Quantity
Catalog number: A19348.06
also known as A19348-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
771-60-8
IUPAC Name
pentafluoroaniline
Molecular Formula
C6H2F5N
InChI Key
NOXLGCOSAFGMDV-UHFFFAOYSA-N
SMILES
NC1=C(F)C(F)=C(F)C(F)=C1F
Appearance (Color)
White to yellow to orange to brown
Form
Fused solid
Assay (GC)
≥96.0%
Melting Point (clear melt)
31.0-40.0?C

Description

2,3,4,5,6-Pentafluoroaniline finds it uses in the preparation of pentafluorophenylammonium triflate, an efficient catalyst for esterification and thioesterification. It is also used in the synthesis of various titanium complexes having two anionic [N, O-] bidentate salicylaldiminato ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluoroaniline finds it uses in the preparation of pentafluorophenylammonium triflate, an efficient catalyst for esterification and thioesterification. It is also used in the synthesis of various titanium complexes having two anionic [N, O-] bidentate salicylaldiminato ligands.

Solubility
Soluble in toluene, and organic solvents. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text