2,3,4,5,6-Pentafluoroaniline finds it uses in the preparation of pentafluorophenylammonium triflate, an efficient catalyst for esterification and thioesterification. It is also used in the synthesis of various titanium complexes having two anionic [N, O-] bidentate salicylaldiminato ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3,4,5,6-Pentafluoroaniline finds it uses in the preparation of pentafluorophenylammonium triflate, an efficient catalyst for esterification and thioesterification. It is also used in the synthesis of various titanium complexes having two anionic [N, O-] bidentate salicylaldiminato ligands.
Solubility
Soluble in toluene, and organic solvents. Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
Funatomi T, et al. Pentafluorophenylammonium triflate (PFPAT): an efficient, practical, and cost-effective catalyst for esterification, thioesterification, transesterification, and macrolactone formation.Green Chem.2006,8(12), 1022-1027.
Parssinen A, et al. Bis (salicylaldiminato) titanium complexes containing bulky imine substituents: Synthesis, characterization and ethene polymerization studies.Eur. J. Inorg. Chem. 2005,2005(11), 2100-2109.