4-Chloro-2-fluoro-1-iodobenzene, 98%, Thermo Scientific Chemicals
4-Chloro-2-fluoro-1-iodobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloro-2-fluoro-1-iodobenzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number A19376.14
also known as A19376-14
Price (USD)/ Each
212.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
212.00
Add to cart
4-Chloro-2-fluoro-1-iodobenzene, 98%, Thermo Scientific Chemicals
Catalog numberA19376.14
Price (USD)/ Each
212.00
-
Add to cart
Chemical Identifiers
CAS6797-79-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale red to red to brown
Refractive Index1.6045-1.6085 @ 20?C
FormLiquid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only