Thermo Scientific Chemicals

cis-3-Hexenyl acetate, 98+%, Thermo Scientific Chemicals

Catalog number: A19380.22
100 g, Each
Thermo Scientific Chemicals

cis-3-Hexenyl acetate, 98+%, Thermo Scientific Chemicals

Catalog number: A19380.22
100 g, Each
Quantity
Catalog number: A19380.22
also known as A19380-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3681-71-8
IUPAC Name
(3Z)-hex-3-en-1-yl acetate
Molecular Formula
C8H14O2
InChI Key
NPFVOOAXDOBMCE-PLNGDYQASA-N
SMILES
CCC=C/CCOC(C)=O
Assay (GC)
≥98.0%
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.4255-1.4285 @ 20°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text