Thermo Scientific Chemicals

2,4,5-Trichlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A19390.22
100 g, Each
Thermo Scientific Chemicals

2,4,5-Trichlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A19390.22
100 g, Each
Quantity
Catalog number: A19390.22
also known as A19390-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15945-07-0
IUPAC Name
2,4,5-trichlorobenzene-1-sulfonyl chloride
Molecular Formula
C6H2Cl4O2S
InChI Key
WNVVRCKTQSCPAC-UHFFFAOYSA-N
SMILES
ClC1=CC(Cl)=C(C=C1Cl)S(Cl)(=O)=O
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Melting Point (clear melt)
64.0-70.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text