Thermo Scientific Chemicals

1,3-Cyclohexadiene, 96%, stab. with 0.1% BHT, Thermo Scientific Chemicals

Catalog number: A19394.14
25 g, Each
Thermo Scientific Chemicals

1,3-Cyclohexadiene, 96%, stab. with 0.1% BHT, Thermo Scientific Chemicals

Catalog number: A19394.14
25 g, Each
Quantity
Catalog number: A19394.14
also known as A19394-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
592-57-4
IUPAC Name
cyclohexa-1,3-diene
Molecular Formula
C6H8
InChI Key
MGNZXYYWBUKAII-UHFFFAOYSA-N
SMILES
C1CC=CC=C1
Appearance (Color)
Clear colorless
Refractive Index
1.4715-1.4765 @ 20*C
Stabilizer
0.1% BHT
Form
Liquid
Assay (GC)
≥95.0%

Description

1,3-Cyclohexadiene is used as a hydrogen donor in transfer hydrogenation. It is used in the conversion to benzene. It is useful in the study of proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Cyclohexadiene is used as a hydrogen donor in transfer hydrogenation. It is used in the conversion to benzene. It is useful in the study of proteomics research.

Solubility
Slightly miscible with methanol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text