It finds its uses as a catalyst to make polyesters, a filler for polypropylene (flame-resistant), and in glass optical waveguides. It also finds ints use in laboratories and metallurgy. it is used as acryl amide monomer light aggregation function of catalysts and can be used for photoelectric material and heat power semiconductor materials; it is also used as analytical reagent, additive and in the preparation of massicot pigment; it is also used as pigment ingredient and used for printing and dyeing industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its uses as a catalyst to make polyesters, a filler for polypropylene (flame-resistant), and in glass optical waveguides. It also finds ints use in laboratories and metallurgy. it is used as acryl amide monomer light aggregation function of catalysts and can be used for photoelectric material and heat power semiconductor materials; it is also used as analytical reagent, additive and in the preparation of massicot pigment; it is also used as pigment ingredient and used for printing and dyeing industry.
Solubility
Soluble in water at 20°C 5g/L. Insoluble in ethanol. Solubility in N,N-DMF is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
- G Ahmed, et al. Electronic properties of PbCl2 and PbBr2 using Compton scattering technique.Appl. Radiat. Isot.2009,67(6), 1050-1056.
- Robin D. Rogers, et al. Structural Chemistry of Poly(ethylene glycol) Complexes of Lead(II) Nitrate and Lead(II) Bromide.Inorg. Chem.,1996,35(24), 6964-6973.