Thermo Scientific Chemicals

4-tert-Butylphenoxyacetyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A19408.18
50 g, Each
Thermo Scientific Chemicals

4-tert-Butylphenoxyacetyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A19408.18
50 g, Each
Quantity
Catalog number: A19408.18
also known as A19408-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
90734-55-7
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Refractive Index
1.5145-1.5185 @ 20?C
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%

Description

4-tert-Butylphenoxyacetyl chloride is used as a reagent and as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylphenoxyacetyl chloride is used as a reagent and as intermediates.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, alcohol, amine, bases, alkali metals, water, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text