Cyclopentadecane, 98%, Thermo Scientific Chemicals
Cyclopentadecane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopentadecane, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number A19416.06
also known as A19416-06
Price (USD)/ Each
159.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
159.00
Add to cart
Cyclopentadecane, 98%, Thermo Scientific Chemicals
Catalog numberA19416.06
Price (USD)/ Each
159.00
-
Add to cart
Chemical Identifiers
CAS295-48-7
IUPAC Namecyclopentadecane
Molecular FormulaC15H30
InChI KeySRONXYPFSAKOGH-UHFFFAOYSA-N
SMILESC1CCCCCCCCCCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)58.0-64.0?C
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only