Thermo Scientific Chemicals

4-Sulfobenzoic acid monopotassium salt, 95%, Thermo Scientific Chemicals

Catalog number: A19432.18
50 g, Each
Thermo Scientific Chemicals

4-Sulfobenzoic acid monopotassium salt, 95%, Thermo Scientific Chemicals

Catalog number: A19432.18
50 g, Each
Quantity
Catalog number: A19432.18
also known as A19432-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5399-63-3
IUPAC Name
potassium 4-sulfobenzoic acid
Molecular Formula
C7H6KO5S
InChI Key
PXRJBUPXKDXDLG-UHFFFAOYSA-N
SMILES
[K+].OC(=O)C1=CC=C(C=C1)S(O)(=O)=O
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Water Content (Karl Fischer Titration)
≤2.0%
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text