4-Sulfobenzoic acid monopotassium salt, 95%, Thermo Scientific Chemicals
4-Sulfobenzoic acid monopotassium salt, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Sulfobenzoic acid monopotassium salt, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number A19432.30
also known as A19432-30
Price (USD)/ Each
189.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
189.00
Add to cart
4-Sulfobenzoic acid monopotassium salt, 95%, Thermo Scientific Chemicals
Catalog numberA19432.30
Price (USD)/ Each
189.00
-
Add to cart
Chemical Identifiers
CAS5399-63-3
IUPAC Namepotassium 4-sulfobenzoic acid
Molecular FormulaC7H6KO5S
InChI KeyPXRJBUPXKDXDLG-UHFFFAOYSA-N
SMILES[K+].OC(=O)C1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Water Content (Karl Fischer Titration)≤2.0%
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only