Thermo Scientific Chemicals

D-(+)-Trehalose dihydrate, 99%

Catalog number: A19434.14
25 g, Each
Thermo Scientific Chemicals

D-(+)-Trehalose dihydrate, 99%

Catalog number: A19434.14
25 g, Each
Quantity
Catalog number: A19434.14
also known as A19434-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6138-23-4
IUPAC Name
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol dihydrate
Molecular Formula
C12H26O13
InChI Key
DPVHGFAJLZWDOC-DJCWUSJTNA-N
SMILES
O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Assay from Supplier's CofA
≥98.5%
Identification (FTIR)
Conforms
Optical Rotation
+178 ± 4? (c=2 in water)
Form
Crystalline powder
Water Content (Karl Fischer Titration)
8.65-10.80% (1.8-2.3 waters)

Description

D-(+)-Trehalose dihydrate is used as a food additive in many processed foods and pharmaceutical excipients. It is widely used as a direct replacement of conventional sweeteners and protects the cells from various environmental stresses. It finds application in the treatment of Huntington's disease and Parkinson's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and hot alcohol.

Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text