2-Isopropylphenyl isothiocyanate, 96%, Thermo Scientific Chemicals
2-Isopropylphenyl isothiocyanate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Isopropylphenyl isothiocyanate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A19448.06
also known as A19448-06
Price (USD)/ Each
85.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
85.50
Add to cart
2-Isopropylphenyl isothiocyanate, 96%, Thermo Scientific Chemicals
Catalog numberA19448.06
Price (USD)/ Each
85.50
-
Add to cart
Chemical Identifiers
CAS36176-31-5
IUPAC Name1-isothiocyanato-2-(propan-2-yl)benzene
Molecular FormulaC10H11NS
InChI KeyQQOOMDSEMQHMIR-UHFFFAOYSA-N
SMILESCC(C)C1=CC=CC=C1N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
FormLiquid
Refractive Index1.6020-1.6080 @ 20?C
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only