4-(2-Furyl)-3-buten-2-one, cis + trans, 98%, Thermo Scientific Chemicals
4-(2-Furyl)-3-buten-2-one, cis + trans, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(2-Furyl)-3-buten-2-one, cis + trans, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A19456.06
also known as A19456-06
Price (USD)/ Each
41.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
41.70
Add to cart
4-(2-Furyl)-3-buten-2-one, cis + trans, 98%, Thermo Scientific Chemicals
Catalog numberA19456.06
Price (USD)/ Each
41.70
-
Add to cart
Chemical Identifiers
CAS623-15-4
IUPAC Name(3E)-4-(furan-2-yl)but-3-en-2-one
Molecular FormulaC8H8O2
InChI KeyGBKGJMYPQZODMI-SNAWJCMRSA-N
SMILESCC(=O)\C=C\C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Appearance (Color)Pale yellow to orange to brown
Melting Point (clear melt)35.0-41.0°C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only