2,6-Dimethylphenyl isocyanate has been used in the preparation of derivatized β-cyclodextrins, used as chiral stationary phase in normal-phase liquid chromatography, tris( 2,6-dimethylphenylimido)methylrhenium (VII), 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dimethylphenyl isocyanate has been used in the preparation of derivatized β-cyclodextrins, used as chiral stationary phase in normal-phase liquid chromatography, tris( 2,6-dimethylphenylimido)methylrhenium (VII), 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
Xuechun Zhang.; Young K. Lee.; James A. Kelley.; Terrence R. Burke. Preparation of Aryl Isothiocyanates via Protected Phenylthiocarbamates and Application to the Synthesis of Caffeic Acid (4-Isothiocyanato)phenyl Ester. J. Org. Chem. 2000, 65, (9), 6237-6240.
Jian Liu.; Raymond J. Patch.; Carsten Schubert.; Mark R. Player. Single-Step Syntheses of 2-Amino-7-chlorothiazolo[5,4-d]pyrimidines: Intermediates for Bivalent Thiazolopyrimidines. J. Org. Chem. 2005, 70, (24), 10194-10197.