Thermo Scientific Chemicals

9-Decen-1-ol, 90+%, Thermo Scientific Chemicals

Catalog number: A19490.14
25 g, Each
Thermo Scientific Chemicals

9-Decen-1-ol, 90+%, Thermo Scientific Chemicals

Catalog number: A19490.14
25 g, Each
Quantity
Catalog number: A19490.14
also known as A19490-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13019-22-2
IUPAC Name
dec-9-en-1-ol
Molecular Formula
C10H20O
InChI Key
QGFSQVPRCWJZQK-UHFFFAOYSA-N
SMILES
OCCCCCCCCC=C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥90.0%
Identification (FTIR)
Conforms
Refractive Index
1.4080-1.4880 @ 20°C

Description

It finds it uses in a wide range of fragrances, especially in fine fragrances and soaps, in floral, rose petal or herbal fragrances. 9-Decen-1-ol is used in the preparation of semifluorinated acids required for the synthesis of poly(styrene-b-semi fluorinated isoprene) block copolymers with -CF2H-terminated side groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds it uses in a wide range of fragrances, especially in fine fragrances and soaps, in floral, rose petal or herbal fragrances. 9-Decen-1-ol is used in the preparation of semifluorinated acids required for the synthesis of poly(styrene-b-semi fluorinated isoprene) block copolymers with -CF2H-terminated side groups.

Solubility
Soluble in water at 20°C 0.16g/L. Soluble in alcohol, dipropylene glycol, paraffin oil.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text