Ethyl isothiocyanate, 97%, Thermo Scientific Chemicals
Ethyl isothiocyanate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl isothiocyanate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number A19492.22
also known as A19492-22
Price (USD)/ Each
138.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
138.00
Add to cart
Ethyl isothiocyanate, 97%, Thermo Scientific Chemicals
Catalog numberA19492.22
Price (USD)/ Each
138.00
-
Add to cart
Chemical Identifiers
CAS542-85-8
IUPAC Nameisothiocyanatoethane
Molecular FormulaC3H5NS
InChI KeyHBNYJWAFDZLWRS-UHFFFAOYSA-N
SMILESCCN=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5100-1.5190 @ 20?C
FormLiquid
CommentMay form slight precipitate on storage.
Appearance (Color)Clear or slightly turbid, colorless to pale yellow
Assay (GC)≥96.0%
View more
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Reacts with water. Soluble in chlorinated and aromatic hydrocarbons. Miscible in ethanol and ether.

Notes
Moisture sensitive. Store away from alcohols, amines, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

General References:

  1. Antonietta Melchini, et al. Enhanced in vitro biological activity of synthetic 2-(2-pyridyl) ethyl isothiocyanate compared to natural 4-(methylsulfinyl) butyl isothiocyanate.Journal of medicinal and pharmaceutical chemistry.2012,55(22), 9682-9692.
  2. Wu H, et al. Antifungal vapor-phase Activity of a Combination of Allyl Isothiocyanate and Ethyl Isothiocyanate Against Botrytis cinerea and Penicillium expansum Infection on Apples.Journal of phytopathology.2011,159(6), 450-455.