N,N,N',N'-Tetramethyl-1,3-propanediamine, 98+%, Thermo Scientific Chemicals
N,N,N',N'-Tetramethyl-1,3-propanediamine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N,N',N'-Tetramethyl-1,3-propanediamine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number A19500.30
also known as A19500-30
Price (USD)/ Each
179.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
179.00
Add to cart
N,N,N',N'-Tetramethyl-1,3-propanediamine, 98+%, Thermo Scientific Chemicals
Catalog numberA19500.30
Price (USD)/ Each
179.00
-
Add to cart
Chemical Identifiers
CAS110-95-2
IUPAC Name[3-(dimethylazaniumyl)propyl]dimethylazanium
Molecular FormulaC7H20N2
InChI KeyDMQSHEKGGUOYJS-UHFFFAOYSA-P
SMILESC[NH+](C)CCC[NH+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Chromatographic PurityConforms
FormLiquid
Cloud pointConforms
Water Content (Karl Fischer Titration)≤0.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only