Thermo Scientific Chemicals

Gold(III) bromide, 99%, Thermo Scientific Chemicals

Catalog number: A19514.03
1 g, Each
Thermo Scientific Chemicals

Gold(III) bromide, 99%, Thermo Scientific Chemicals

Catalog number: A19514.03
1 g, Each
Quantity
Catalog number: A19514.03
also known as A19514-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10294-28-7
Appearance (Color)
Red/brown or grey to black
Assay (unspecified)
(based on Au Elemental Analysis) : >98.5 to <101.5%
Elemental Analysis
(Gold, Au) : 44.4-45.8%
Form
Powder

Description

Gold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text