Ethyl 2-methyl-4-pentenoate, 98%, Thermo Scientific Chemicals
Ethyl 2-methyl-4-pentenoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-methyl-4-pentenoate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number A19526.22
also known as A19526-22
Price (USD)/ Each
209.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
209.00
Add to cart
Ethyl 2-methyl-4-pentenoate, 98%, Thermo Scientific Chemicals
Catalog numberA19526.22
Price (USD)/ Each
209.00
-
Add to cart
Chemical Identifiers
CAS53399-81-8
IUPAC Nameethyl 2-methylpent-4-enoate
Molecular FormulaC8H14O2
InChI KeyBDBGKYIBDXAVMX-UHFFFAOYNA-N
SMILESCCOC(=O)C(C)CC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4145-1.4185 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only