Thermo Scientific Chemicals

2-Acetyl-1-naphthol, 98+%, Thermo Scientific Chemicals

Catalog number: A19532.14
25 g, Each
Thermo Scientific Chemicals

2-Acetyl-1-naphthol, 98+%, Thermo Scientific Chemicals

Catalog number: A19532.14
25 g, Each
Quantity
Catalog number: A19532.14
also known as A19532-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
711-79-5
IUPAC Name
1-(1-hydroxynaphthalen-2-yl)ethan-1-one
Molecular Formula
C12H10O2
InChI Key
JBGJVMVWYWUVOW-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C2C=CC=CC2=C1O
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
95.0-102.0°C
Appearance (Color)
Yellow
Assay (GC)
≥98.0%

Description

2-acetyl-1-naphthol and 1-acetyl-2-naphthol with triphosgene gave naphtho[1,2-e]-1,3-oxazines, naphtho[2,1-e]-1,3-oxazines or their spiro dimers depending on the molar ratio of triphosgene used for the cyclization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-acetyl-1-naphthol and 1-acetyl-2-naphthol with triphosgene gave naphtho[1,2-e]-1,3-oxazines, naphtho[2,1-e]-1,3-oxazines or their spiro dimers depending on the molar ratio of triphosgene used for the cyclization.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text