Diethylbenzene, mixture of isomers, Thermo Scientific Chemicals
Diethylbenzene, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethylbenzene, mixture of isomers, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2500 mL
Catalog number A19548.AP
also known as A19548-AP
Price (USD)/ Each
40.40
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
40.40
Add to cart
Diethylbenzene, mixture of isomers, Thermo Scientific Chemicals
Catalog numberA19548.AP
Price (USD)/ Each
40.40
-
Add to cart
Chemical Identifiers
CAS25340-17-4
IUPAC Name1,2-diethylbenzene
Molecular FormulaC10H14
InChI KeyKVNYFPKFSJIPBJ-UHFFFAOYSA-N
SMILESCCC1=CC=CC=C1CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4925-1.4985 @ 20°C
FormLiquid
Assay (GC)≥95.0% (sum of isomers)
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only