Benzyl cinnamate, 99%, Thermo Scientific Chemicals
Benzyl cinnamate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyl cinnamate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A19550.22
also known as A19550-22
Price (USD)/ Each
40.90
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
40.90
Add to cart
Benzyl cinnamate, 99%, Thermo Scientific Chemicals
Catalog numberA19550.22
Price (USD)/ Each
40.90
-
Add to cart
Chemical Identifiers
CAS103-41-3
IUPAC Namebenzyl (2E)-3-phenylprop-2-enoate
Molecular FormulaC16H14O2
InChI KeyNGHOLYJTSCBCGC-VAWYXSNFSA-N
SMILESO=C(OCC1=CC=CC=C1)\C=C\C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormFused solid or liquid as melt
Melting Point (clear melt)31.5-37.5?C
Assay (GC)≥98.5%
Appearance (Color)White to very pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only