Thermo Scientific Chemicals

3-(Trifluoromethoxy)aniline, 98%, Thermo Scientific Chemicals

Catalog number: A19592.06
5 g, Each
Thermo Scientific Chemicals

3-(Trifluoromethoxy)aniline, 98%, Thermo Scientific Chemicals

Catalog number: A19592.06
5 g, Each
Quantity
Catalog number: A19592.06
also known as A19592-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1535-73-5
IUPAC Name
3-(trifluoromethoxy)aniline
Molecular Formula
C7H6F3NO
InChI Key
SADHVOSOZBAAGL-UHFFFAOYSA-N
SMILES
NC1=CC=CC(OC(F)(F)F)=C1
Appearance (Color)
Clear pale yellow to pale brown
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4640-1.4680 @ 20?C

Description

3-(Trifluoromethoxy)aniline may be used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Trifluoromethoxy)aniline may be used in chemical synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text