Thermo Scientific Chemicals

1-Bromo-3-iodobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A19596.14
25 g, Each
Thermo Scientific Chemicals

1-Bromo-3-iodobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A19596.14
25 g, Each
Quantity
Catalog number: A19596.14
also known as A19596-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
591-18-4
IUPAC Name
1-bromo-3-iodobenzene
Molecular Formula
C6H4BrI
InChI Key
CTPUUDQIXKUAMO-UHFFFAOYSA-N
SMILES
BrC1=CC=CC(I)=C1
Appearance (Color)
Clear colorless to yellow to brown
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.6595-1.6635 @ 20?C

Description

1-Bromo-3-iodobenzene is used in the preparation of 3'-bromo-2,3,4,5,6-pentaethyl-biphenyl by reacting with hex-3-yne. It is also used to prepare 1-(3'-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene and 1-(3'-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene. Further, it is involved in the preparation of oxygen-tethered 1,6-enynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-iodobenzene is used in the preparation of 3′-bromo-2,3,4,5,6-pentaethyl-biphenyl by reacting with hex-3-yne. It is also used to prepare 1-(3′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene and 1-(3′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene. Further, it is involved in the preparation of oxygen-tethered 1,6-enynes.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong acids. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text