tert-Nitrobutane, 98+%, Thermo Scientific Chemicals
tert-Nitrobutane, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Nitrobutane, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A19608.06
also known as A19608-06
Price (USD)/ Each
41.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
41.90
Add to cart
tert-Nitrobutane, 98+%, Thermo Scientific Chemicals
Catalog numberA19608.06
Price (USD)/ Each
41.90
-
Add to cart
Chemical Identifiers
CAS594-70-7
IUPAC Name2-methyl-2-nitropropane
Molecular FormulaC4H9NO2
InChI KeyAIMREYQYBFBEGQ-UHFFFAOYSA-N
SMILESCC(C)(C)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
FormFused solid or clear liquid as melt
Assay (GC)≥98.0%
Refractive Index1.3970-1.4015 @ 30?C
It is used in the cyclic voltammetric study of tert-nitrobutane reduction in acetonitrile at mercury and platinum electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the cyclic voltammetric study of tert-nitrobutane reduction in acetonitrile at mercury and platinum electrodes.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Sumana Sengupta, et. al. Photodissociation dynamics of 2-nitropropane and 2-methyl-2-nitropropane at 248 and 193 nm.J Phys Chem A.2008,112(49), 12572-12581.
  2. Joubert E, et al. N2O formation during organic nitro compounds' hydrolysis on platinum based catalysts.Appl. Catal. B.1999,23(2), 83-87.