Thermo Scientific Chemicals

Arachidic acid, 98%, Thermo Scientific Chemicals

Catalog number: A19616.14
25 g, Each
Thermo Scientific Chemicals

Arachidic acid, 98%, Thermo Scientific Chemicals

Catalog number: A19616.14
25 g, Each
Quantity
Catalog number: A19616.14
also known as A19616-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
506-30-9
IUPAC Name
icosanoic acid
Molecular Formula
C20H40O2
InChI Key
VKOBVWXKNCXXDE-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCC(O)=O
Appearance (Color)
White or translucent
Form
Crystals, crystalline powder or flakes
Assay (GC)
≥97.5%
Melting Point (clear melt)
73-79?C

Description

Arachidic acid is used in the manufacture of pharmaceuticals, soaps, cosmetics, and food packaging because of its surfactant-like properties. Arachidic acid is a saturated fatty acid with a 20 carbon chain. Diets rich in saturated fats like arachidic acid are associated with increased levels of serum low density lipoproteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Arachidic acid is used in the manufacture of pharmaceuticals, soaps, cosmetics, and food packaging because of its surfactant-like properties. Arachidic acid is a saturated fatty acid with a 20 carbon chain. Diets rich in saturated fats like arachidic acid are associated with increased levels of serum low density lipoproteins.

Solubility
Soluble in chloroform, ether, ethanol, and DMF. Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text